Assay Details
| Gene Symbol: Neil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028347.2 | NP_082623.1 | 4 - 5 | 717 | 70 | |
| GenBank mRNA | AB079069.1 | - | 4 - 5 | 717 | 70 | |
| AK013322.1 | - | 5 - 6 | 788 | 70 | ||
| AK155405.1 | - | 6 - 7 | 593 | 70 | ||
| AK186023.1 | - | 3 - 4 | 278 | 70 | ||
| BC034873.1 | - | 5 - 6 | 962 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Neil1
Genomic Map