Assay Details
Gene Symbol: Mapk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015806.5 | NP_056621.4 | 3 - 4 | 1471 | 67 | |
NM_027418.2 | NP_081694.1 | 3 - 4 | 1475 | 67 | ||
GenBank mRNA | AF132850.1 | - | 2 - 3 | 1204 | 67 | |
BC006778.1 | - | 2 - 3 | 685 | 67 | ||
BC037060.1 | - | 2 - 3 | 1209 | 67 | ||
BC048779.1 | - | 2 - 3 | 973 | 6315375 | 67 | |
BC052420.1 | - | 2 - 3 | 720 | 5702905 | 67 | |
BC100385.1 | - | 3 - 4 | 1453 | 30732053 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
