Assay Details
Gene Symbol: Arih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019927.2 | NP_064311.2 | 11 - 12 | 1235 | 104 | |
GenBank mRNA | AK087802.1 | - | 1 - 2 | 67 | 104 | |
BC038034.1 | - | 11 - 12 | 1155 | 4035861 | 104 | |
BC057680.1 | - | 11 - 12 | 1475 | 3498094 | 104 | |
CK633971.1 | - | 2 - 3 | 130 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
