Assay Details
Gene Symbol: Arih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019927.2 | NP_064311.2 | 4 - 5 | 697 | 120 | |
GenBank mRNA | AF361001.1 | - | 4 - 5 | 411 | 120 | |
AJ130977.1 | - | 4 - 5 | 394 | 120 | ||
BC038034.1 | - | 4 - 5 | 617 | 4035861 | 120 | |
BC057680.1 | - | 4 - 5 | 937 | 3498094 | 120 | |
CA328699.1 | - | 4 - 5 | 644 | 120 | ||
CV555932.1 | - | 4 - 5 | 352 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
