Assay Details
Gene Symbol: Ranbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011239.2 | NP_035369.2 | 1 - 2 | 150 | 93 | |
GenBank mRNA | AK002989.1 | - | 1 - 2 | 116 | 93 | |
AK008276.1 | - | 1 - 2 | 113 | 93 | ||
AK088781.1 | - | 1 - 2 | 138 | 93 | ||
AK151167.1 | - | 1 - 2 | 128 | 93 | ||
AK153074.1 | - | 1 - 2 | 132 | 93 | ||
AK166642.1 | - | 1 - 2 | 152 | 93 | ||
AK167053.1 | - | 1 - 2 | 134 | 93 | ||
BC061140.1 | - | 1 - 2 | 169 | 6775220 | 93 | |
X56045.1 | - | 1 - 2 | 155 | 92 | ||
X56046.1 | - | 1 - 2 | 155 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK002989.1, AK008276.1, AK088781.1, AK151167.1, AK153074.1, AK166642.1, AK167053.1, BC061140.1, NM_011239.2, X56045.1, X56046.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
