Assay Details
| Gene Symbol: Sema7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011352.2 | NP_035482.1 | 5 - 6 | 557 | 68 | |
| GenBank mRNA | AB017532.1 | - | 5 - 6 | 546 | 68 | |
| AF030699.1 | - | 5 - 6 | 553 | 68 | ||
| AF176670.1 | - | 5 - 6 | 546 | 68 | ||
| AK156123.1 | - | 5 - 6 | 580 | 68 | ||
| BC057875.1 | - | 5 - 6 | 557 | 5319572 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sema7a
Genomic Map