Assay Details
| Gene Symbol: Dennd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162917.1 | NP_001156389.1 | 32 - 33 | 5882 | 82 | |
| GenBank mRNA | AK035923.1 | - | 10 - 11 | 1980 | 82 | |
| AK046940.1 | - | 1 - 2 | 190 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dennd4a
Genomic Map