Assay Details
| Gene Symbol: Dennd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162917.1 | NP_001156389.1 | 24 - 25 | 4720 | 59 | |
| GenBank mRNA | AK035923.1 | - | 2 - 3 | 818 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dennd4a
Genomic Map