Assay Details
| Gene Symbol: Pld3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011116.1 | NP_035246.1 | 1 - 2 | 110 | 123 | |
| GenBank mRNA | AF026124.1 | - | 1 - 2 | 110 | 123 | |
| BC068170.1 | - | 1 - 2 | 107 | 123 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF026124.1, NM_011116.1 transcripts.
Set Membership: |
|

Pld3
Genomic Map