Assay Details
| Gene Symbol: Carm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021531.6 | NP_067506.2 | 11 - 12 | 1455 | 68 | |
| NM_153141.1 | NP_694781.1 | 11 - 12 | 1455 | 68 | ||
| GenBank mRNA | AF117887.2 | - | 11 - 12 | 1365 | 68 | |
| AK158242.1 | - | 11 - 12 | 1420 | 68 | ||
| AK158757.1 | - | 11 - 12 | 1295 | 68 | ||
| BC003964.1 | - | 6 - 7 | 562 | 68 | ||
| BC008263.1 | - | 1 - 2 | 61 | 68 | ||
| BC036974.1 | - | 11 - 12 | 1439 | 4935077 | 68 | |
| BC071267.1 | - | 12 - 13 | 1571 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Carm1
Genomic Map