Assay Details
| Gene Symbol: Carm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021531.6 | NP_067506.2 | 14 - 15 | 1728 | 74 | |
| GenBank mRNA | AF117887.2 | - | 14 - 15 | 1638 | 74 | |
| AK158242.1 | - | 14 - 15 | 1693 | 74 | ||
| AK158757.1 | - | 14 - 15 | 1568 | 74 | ||
| AK189449.1 | - | 1 - 2 | 37 | 74 | ||
| BC003964.1 | - | 9 - 10 | 835 | 74 | ||
| BC008263.1 | - | 4 - 5 | 334 | 74 | ||
| BC026427.1 | - | 4 - 5 | 294 | 74 | ||
| BC071267.1 | - | 15 - 16 | 1844 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Carm1
Genomic Map