Assay Details
| Gene Symbol: Shc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024539.1 | NP_001019710.1 | 10 - 11 | 1348 | 63 | |
| GenBank mRNA | AK032878.1 | - | 11 - 12 | 1033 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027077 | BC059866.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027077 | NM_001024539.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P169714 | AK032878.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P169714 | BC059866.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P169714 | NM_001024539.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P169714 | AK032878.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P169714 | BC059866.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P169714 | NM_001024539.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781664 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726489 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 56221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 217727 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327099 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538461 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446129 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148729 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861700 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52302 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110695 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138058 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027077 | BC059866.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027077 | NM_001024539.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P169714 | AK032878.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P169714 | BC059866.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P169714 | NM_001024539.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P169714 | AK032878.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P169714 | BC059866.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P169714 | NM_001024539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P169714 | AK032878.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P169714 | BC059866.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P169714 | NM_001024539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P169714 | AK032878.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P169714 | BC059866.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P169714 | NM_001024539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104108 | BC059866.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104108 | NM_001024539.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08768103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08265848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08361590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08768098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08666528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08361598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08768090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08265837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08265828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08588047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08223524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08361592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08265829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08768093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08592775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08223521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08265847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277986 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10370595 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370591 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370589 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370588 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370598 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370592 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370594 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370597 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370593 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370602 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370590 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370599 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370601 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370596 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441356_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437944_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438598_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10370587 | - | - |
| Mouse Genome 430 2.0 Array | 1437944_at | - | - |
| Mouse Genome 430 2.0 Array | 1441356_at | - | - |
| Mouse Genome 430 2.0 Array | 1441356_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435598_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435598_at | - | - |
| Mouse Genome 430 2.0 Array | 1437944_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438598_at | - | - |
| Mouse Genome 430 2.0 Array | 1438598_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216148_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216148_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000018040.mm.1 | NM_001024539.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018040.mm.1 | NM_001024539.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018040.mm.1 | TC1000002472.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438598_at | - | - |
| Mouse Expression Set 430 | 1437944_PM_at | - | - |
| Mouse Expression Set 430 | 1438598_PM_at | - | - |
| Mouse Expression Set 430 | 1441356_PM_at | - | - |
| Mouse Expression Set 430 | 1441356_at | - | - |
| Mouse Expression Set 430 | 1437944_at | - | - |
| Murine Genome U74v2 Set | 138008_PM_r_at | - | - |
| Murine Genome U74v2 Set | 138008_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438598_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441356_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437944_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438598_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437944_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441356_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 463281 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1940725 | - | - |

Shc2
Genomic Map