Assay Details
| Gene Symbol: Shc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024539.1 | NP_001019710.1 | 11 - 12 | 1663 | 98 | |
| GenBank mRNA | AK032878.1 | - | 12 - 13 | 1348 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shc2
Genomic Map