Assay Details
| Gene Symbol: Shc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024539.1 | NP_001019710.1 | 8 - 9 | 1115 | 62 | |
| GenBank mRNA | AK032878.1 | - | 9 - 10 | 800 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shc2
Genomic Map