Assay Details
| Gene Symbol: Slc17a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276452.1 | NP_001263381.1 | 5 - 6 | 701 | 136 | |
| NM_172773.3 | NP_766361.1 | 5 - 6 | 779 | 136 | ||
| GenBank mRNA | AK029102.1 | - | 5 - 6 | 762 | 136 | |
| AK169868.1 | - | 5 - 6 | 728 | 136 | ||
| BC058785.1 | - | 5 - 6 | 671 | 6309785 | 136 | |
More Information
Set Membership: |
|

Slc17a5
Genomic Map