Assay Details
Gene Symbol: Olfr860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146528.2 | NP_666739.1 | 2 | 864 | 139 | |
GenBank mRNA | BC120862.1 | - | 1 | 695 | 40129785 | 139 |
BC137831.1 | - | 1 | 695 | 8860850 | 139 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
