Assay Details
| Gene Symbol: 2900052N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015605.1 | 2 - 3 | 1558 | 139 | ||
| GenBank mRNA | AK049847.1 | - | 2 - 3 | 1558 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

2900052N01Rik
Genomic Map