Assay Details
| Gene Symbol: Filip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081243.1 | NP_001074712.1 | 3 - 4 | 889 | 87 | |
| GenBank mRNA | AK017709.1 | - | 3 - 4 | 986 | 87 | |
| BC030632.1 | - | 3 - 4 | 534 | 87 | ||
| BC125581.1 | - | 3 - 4 | 734 | 40130096 | 87 | |
| BC131965.1 | - | 3 - 4 | 734 | 40130242 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Filip1
Genomic Map