Assay Details
| Gene Symbol: 4930407I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019573.1 | - | 2 - 3 | 475 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700023F02 gene | 1700023F02Rik | AV205046 | -22.1kb | Anti-sense | Chr. 10 - 66120609 - 66124064 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1110002J07 gene | 1110002J07Rik | 755.8kb | Anti-sense | Chr. 10 - 66912191 - 66920616 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930407I19Rik
Genomic Map