Assay Details
| Gene Symbol: Tle2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019725.2 | NP_062699.1 | 4 - 5 | 684 | 87 | |
| GenBank mRNA | AF145958.2 | - | 4 - 5 | 439 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tle2
Genomic Map