Assay Details
Gene Symbol: Phxr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028271.1 | 1 | 1225 | 74 | ||
GenBank mRNA | X12806.1 | - | 1 | 1237 | 74 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077088 | X12806.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077088 | NR_028271.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077088 | BC107288.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077088 | BC107289.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077088 | BC127147.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439836 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155078 | - | - |
Whole Mouse Genome G4122A | A_51_P483215 | X12806.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P483215 | NR_028271.1 | mouse_transcriptome |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311268 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311209 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311202 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07842219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07644356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311200 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07612547 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311249 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07842221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311190 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311165 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311166 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311192 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311197 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311218 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07959233 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311186 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311225 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311238 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311260 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311176 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311258 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311195 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311188 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07862560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311163 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311217 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07745292 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311251 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311169 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311227 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311158 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311206 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311180 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311233 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311215 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07504570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311171 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26311155 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10583206 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583204 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583205 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1422272_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10583203 | - | - |
Mouse Genome 430 2.0 Array | 1422272_at | - | - |
Mouse Genome 430 2.0 Array | 1422272_PM_at | - | - |
Mouse Expression Set 430 | 1422272_PM_at | - | - |
Mouse Expression Set 430 | 1422272_at | - | - |
Murine Genome U74v2 Set | 101700_at | - | - |
Murine Genome U74v2 Set | 101700_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422272_at | - | - |
Mouse Genome 430A 2.0 Array | 1422272_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422272_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422272_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 470803 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1770161 | - | - |
