Assay Details
Gene Symbol: Sept10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024910.3 | NP_001020081.2 | 2 - 3 | 572 | 143 | |
NM_001024911.2 | NP_001020082.1 | 2 - 3 | 572 | 143 | ||
GenBank mRNA | AK076569.1 | - | 2 - 3 | 573 | 143 | |
BC053752.1 | - | 2 - 3 | 186 | 6506619 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
