Assay Details
| Gene Symbol: Hnrnpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177301.5 | NP_796275.3 | 3 - 4 | 639 | 65 | |
| GenBank mRNA | AK044748.1 | - | 3 - 4 | 415 | 65 | |
| AK085906.1 | - | 3 - 4 | 590 | 65 | ||
| AK144684.1 | - | 3 - 4 | 593 | 65 | ||
| BC027206.1 | - | 3 - 4 | 616 | 4218471 | 65 | |
| BC099683.1 | - | 3 - 4 | 634 | 6530863 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpl
Genomic Map