Assay Details
| Gene Symbol: Hnrnpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177301.5 | NP_796275.3 | 7 - 8 | 962 | 105 | |
| GenBank mRNA | BC027206.1 | - | 7 - 8 | 939 | 4218471 | 105 |
| BC099683.1 | - | 7 - 8 | 957 | 6530863 | 105 | |
| CV561640.1 | - | 2 - 3 | 95 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpl
Genomic Map