Assay Details
| Gene Symbol: Hnrnpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177301.5 | NP_796275.3 | 8 - 9 | 1240 | 69 | |
| GenBank mRNA | AB009392.1 | - | 1 - 2 | 59 | 69 | |
| AK144684.1 | - | 7 - 8 | 1305 | 69 | ||
| BC027206.1 | - | 8 - 9 | 1217 | 4218471 | 69 | |
| BC030461.1 | - | 1 - 2 | 83 | 69 | ||
| BC099683.1 | - | 8 - 9 | 1235 | 6530863 | 69 | |
| CV561640.1 | - | 3 - 4 | 373 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrnpl
Genomic Map