Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700023209.mm.1 |
TC0700004640.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023209.mm.1 |
NM_138676.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023209.mm.1 |
NM_138676.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023217.mm.1 |
TC0700004640.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023217.mm.1 |
NM_138676.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023217.mm.1 |
NM_138676.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023214.mm.1 |
TC0700004640.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023214.mm.1 |
NM_138676.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023214.mm.1 |
NM_138676.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023215.mm.1 |
TC0700004640.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023215.mm.1 |
NM_138676.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023215.mm.1 |
NM_138676.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023206.mm.1 |
TC0700004640.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023206.mm.1 |
NM_138676.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023206.mm.1 |
NM_138676.2 |
Refseq |