Assay Details
Gene Symbol: Dnajb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK038149.1 | - | 7 - 8 | 1124 | 83 | |
AK172485.1 | - | 8 - 9 | 1211 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
