Assay Details
| Gene Symbol: Rhobtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252636.1 | NP_001239565.1 | 1 - 2 | 64 | 89 | |
| GenBank mRNA | AK005770.1 | - | 1 - 2 | 64 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005770.1, NM_001252636.1 transcripts.
Set Membership: |
|

Rhobtb1
Genomic Map