Assay Details
Gene Symbol: Ascc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199187.2 | NP_001186116.1 | 1 - 2 | 85 | 72 | |
GenBank mRNA | AK007519.1 | - | 1 - 2 | 82 | 72 | |
AK139944.1 | - | 1 - 2 | 85 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK007519.1, AK139944.1, NM_001199187.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
