Assay Details
| Gene Symbol: Nde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114085.1 | NP_001107557.1 | 3 - 4 | 366 | 62 | |
| NM_001285503.1 | NP_001272432.1 | 3 - 4 | 366 | 62 | ||
| NM_023317.2 | NP_075806.2 | 3 - 4 | 366 | 62 | ||
| GenBank mRNA | AF322073.1 | - | 2 - 3 | 273 | 62 | |
| AK012830.1 | - | 3 - 4 | 296 | 62 | ||
| AK012861.1 | - | 3 - 4 | 316 | 62 | ||
| AK037289.1 | - | 3 - 4 | 309 | 62 | ||
| AK146982.1 | - | 3 - 4 | 296 | 62 | ||
| AK150826.1 | - | 3 - 4 | 356 | 62 | ||
| AK169743.1 | - | 3 - 4 | 296 | 62 | ||
| BC023267.1 | - | 3 - 4 | 345 | 3672565 | 62 | |
More Information
Set Membership: |
|

Nde1
Genomic Map