Assay Details
| Gene Symbol: Rora | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289916.1 | NP_001276845.1 | 8 - 9 | 1305 | 110 | |
| NM_013646.2 | NP_038674.1 | 9 - 10 | 1314 | 110 | ||
| GenBank mRNA | AK043990.1 | - | 5 - 6 | 674 | 110 | |
| AK149294.1 | - | 8 - 9 | 1195 | 110 | ||
| AK155827.1 | - | 8 - 9 | 1256 | 110 | ||
| BC003757.1 | - | 8 - 9 | 1254 | 3592667 | 110 | |
| S82720.1 | - | 8 - 9 | 1228 | 110 | ||
| U53228.1 | - | 9 - 10 | 1298 | 110 | ||
| Z82994.1 | - | 8 - 9 | 1293 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rora
Genomic Map