Assay Details
| Gene Symbol: Tnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110147.1 | NP_001103617.1 | 10 - 11 | 1939 | 118 | |
| NM_001289443.1 | NP_001276372.1 | 2 - 3 | 326 | 118 | ||
| NM_016788.3 | NP_058068.2 | 10 - 11 | 1939 | 118 | ||
| GenBank mRNA | AF037260.1 | - | 10 - 11 | 1722 | 118 | |
| AK087965.1 | - | 7 - 8 | 1096 | 118 | ||
| BC031168.1 | - | 2 - 3 | 333 | 4984168 | 118 | |
| BC052421.1 | - | 10 - 11 | 1939 | 6400209 | 118 | |
| DQ666696.1 | - | 10 - 11 | 1750 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC031168.1, NM_001289443.1 transcripts.
Set Membership: |
|

Tnk2
Genomic Map