Assay Details
| Gene Symbol: Tnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016788.3 | NP_058068.2 | 13 - 14 | 3427 | 55 | |
| GenBank mRNA | AF037260.1 | - | 13 - 14 | 3210 | 55 | |
| AK087965.1 | - | 9 - 10 | 2539 | 55 | ||
| BC031168.1 | - | 4 - 5 | 1776 | 4984168 | 55 | |
| BC034265.1 | - | 2 - 3 | 202 | 55 | ||
| DQ666696.1 | - | 13 - 14 | 3238 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnk2
Genomic Map