Assay Details
| Gene Symbol: Idh3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029573.2 | NP_083849.1 | 3 - 4 | 179 | 294 | |
| GenBank mRNA | AK010065.1 | - | 3 - 4 | 203 | 294 | |
| AK032787.1 | - | 3 - 4 | 203 | 294 | ||
| AK047951.1 | - | 3 - 4 | 201 | 294 | ||
| AK150618.1 | - | 3 - 4 | 210 | 294 | ||
| AK151304.1 | - | 3 - 4 | 204 | 294 | ||
| AK152353.1 | - | 3 - 4 | 213 | 294 | ||
| AK153459.1 | - | 3 - 4 | 204 | 294 | ||
| AK158646.1 | - | 3 - 4 | 357 | 294 | ||
| AK159051.1 | - | 3 - 4 | 276 | 294 | ||
| AK167822.1 | - | 3 - 4 | 210 | 294 | ||
| AK168034.1 | - | 3 - 4 | 201 | 294 | ||
| AK168049.1 | - | 3 - 4 | 204 | 294 | ||
| AK168149.1 | - | 3 - 4 | 210 | 294 | ||
| AK169152.1 | - | 3 - 4 | 177 | 294 | ||
| BC034273.1 | - | 3 - 4 | 169 | 4166193 | 294 | |
| BC049956.1 | - | 3 - 4 | 179 | 4212315 | 294 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC034273.1 transcripts.
Set Membership: |
|

Idh3a
Genomic Map