Assay Details
| Gene Symbol: Rab27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301230.1 | NP_001288159.1 | 1 - 2 | 202 | 60 | |
| GenBank mRNA | BC008173.1 | - | 1 - 2 | 159 | 3710213 | 60 |
| BY151058.1 | - | 1 - 2 | 169 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC008173.1, NM_001301230.1 transcripts.
Set Membership: |
|

Rab27a
Genomic Map