Assay Details
| Gene Symbol: Pde4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089349.1 | - | 1 - 2 | 450 | 73 | |
| AK170579.1 | - | 1 - 2 | 450 | 73 | ||
| BC027224.1 | - | 1 - 2 | 213 | 73 | ||
More Information
Set Membership: |
|

Pde4a
Genomic Map