Assay Details
| Gene Symbol: Mre11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK146234.1 | - | 9 - 10 | 1179 | 92 | |
| AK146370.1 | - | 9 - 10 | 1179 | 92 | ||
| AK168180.1 | - | 9 - 10 | 1179 | 92 | ||
| U60318.1 | - | 9 - 10 | 1181 | 92 | ||
More Information
Set Membership: |
|

Mre11a
Genomic Map