Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900017874.mm.1 |
NM_138955.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017874.mm.1 |
NM_138955.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017874.mm.1 |
TC0900002153.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017873.mm.1 |
NM_138955.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017873.mm.1 |
NM_138955.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017873.mm.1 |
TC0900002153.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017871.mm.1 |
NM_138955.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017871.mm.1 |
NM_138955.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017871.mm.1 |
TC0900002153.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017872.mm.1 |
NM_138955.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017872.mm.1 |
NM_138955.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017872.mm.1 |
TC0900002153.mm.1 |
MTA TC |