Assay Details
| Gene Symbol: Il10ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008348.2 | NP_032374.1 | 4 - 5 | 619 | 100 | |
| GenBank mRNA | AK137663.1 | - | 4 - 5 | 620 | 100 | |
| AK149680.1 | - | 4 - 5 | 614 | 100 | ||
| AK149899.1 | - | 3 - 4 | 502 | 100 | ||
| AK150022.1 | - | 4 - 5 | 616 | 100 | ||
| AK150524.1 | - | 4 - 5 | 623 | 100 | ||
| AK153120.1 | - | 4 - 5 | 623 | 100 | ||
| AK159566.1 | - | 4 - 5 | 616 | 100 | ||
| L12120.1 | - | 4 - 5 | 624 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il10ra
Genomic Map