Assay Details
Gene Symbol: Calml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102468.1 | NP_001095938.1 | 2 - 3 | 89 | 80 | |
GenBank mRNA | BY097052.1 | - | 2 - 3 | 101 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001102468.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
