Assay Details
| Gene Symbol: Hmbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013551.2 | NP_038579.2 | 2 - 3 | 272 | 94 | |
| GenBank mRNA | AK151020.1 | - | 2 - 3 | 238 | 94 | |
| AK153884.1 | - | 2 - 3 | 205 | 94 | ||
| AK166734.1 | - | 2 - 3 | 170 | 94 | ||
| AK167702.1 | - | 2 - 3 | 231 | 94 | ||
| BC003861.1 | - | 2 - 3 | 232 | 3487642 | 94 | |
| BY786671.1 | - | 2 - 3 | 273 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hmbs
Genomic Map