Assay Details
Gene Symbol: Sdhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025848.3 | NP_080124.1 | 3 - 4 | 372 | 84 | |
GenBank mRNA | AK013503.1 | - | 3 - 4 | 337 | 84 | |
AK013962.2 | - | 3 - 4 | 340 | 84 | ||
AK135970.1 | - | 3 - 4 | 332 | 84 | ||
BC145731.1 | - | 3 - 4 | 332 | 40131034 | 84 | |
BC145733.1 | - | 3 - 4 | 332 | 40131036 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504535 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504536 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260871 | AK013962.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260871 | AK135970.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260871 | AK013503.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260871 | BF662071.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260871 | NM_025848.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260871 | AK013962.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260871 | AK135970.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260871 | AK013503.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260871 | BF662071.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P260871 | NM_025848.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98707 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 214338 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47865 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672547 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113258 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 6475 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131083 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260871 | AK013962.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260871 | AK135970.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260871 | AK013503.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260871 | BF662071.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260871 | NM_025848.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260871 | AK013962.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260871 | AK135970.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260871 | AK013503.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260871 | BF662071.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P260871 | NM_025848.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260871 | AK013962.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260871 | AK135970.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260871 | AK013503.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260871 | BF662071.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260871 | NM_025848.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260871 | AK013962.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260871 | AK135970.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260871 | AK013503.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260871 | BF662071.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260871 | NM_025848.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504522 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504527 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504519 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504530 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504509 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504531 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504529 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504524 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504533 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504525 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504512 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504521 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504511 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504514 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504516 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504517 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504536 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504510 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504513 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131083 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06785286 | - | - |
Whole Mouse Genome G4122A | A_51_P260871 | AK013962.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260871 | AK135970.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260871 | AK013503.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260871 | BF662071.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260871 | NM_025848.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260871 | AK013962.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260871 | AK135970.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260871 | AK013503.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260871 | BF662071.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P260871 | NM_025848.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123508 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131083 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08035969 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08035978 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07683769 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07683760 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504536 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504494 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07910969 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504529 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504471 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504509 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504512 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07532637 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504469 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504497 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504527 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08035978 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07683769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504501 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504470 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504514 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07689181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504525 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504518 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504519 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08078735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504493 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504536 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504475 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07732769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504521 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26504510 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07772950 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10593358 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10593360 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10593357 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10593359 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1437489_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428235_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10593356 | - | - |
Mouse Genome 430 2.0 Array | 1428235_at | - | - |
Mouse Genome 430 2.0 Array | 1428235_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1437489_x_at | - | - |
Mouse Genome 430 2.0 Array | 1437489_PM_x_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66925_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66925_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900018881.mm.1 | NM_025848.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900018881.mm.1 | NM_025848.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900018881.mm.1 | TC0900002271.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900018882.mm.1 | NM_025848.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900018882.mm.1 | NM_025848.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900018882.mm.1 | TC0900002271.mm.1 | MTA TC |
Mouse Expression Set 430 | 1428235_PM_at | - | - |
Mouse Expression Set 430 | 1437489_PM_x_at | - | - |
Mouse Expression Set 430 | 1437489_x_at | - | - |
Mouse Expression Set 430 | 1428235_at | - | - |
Murine Genome U74v2 Set | 92574_PM_at | - | - |
Murine Genome U74v2 Set | 92574_at | - | - |
Murine Genome U74v2 Set | 169663_at | - | - |
Murine Genome U74v2 Set | 169663_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1428235_at | - | - |
Mouse Genome 430A 2.0 Array | 1437489_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1437489_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1428235_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1437489_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1437489_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428235_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428235_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 557782 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2570128 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002570128 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239143 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1239143 | - | - |
mouse-6 v1.1 (Illumina) | scl35879.4_19-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2570128 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl35879.4_19-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2570128 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1239143 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239143 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl35879.4_19-S | - | - |
