Assay Details
| Gene Symbol: Igdcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008988.2 | NP_033014.1 | 10 - 11 | 2010 | 66 | |
| GenBank mRNA | AF026465.1 | - | 10 - 11 | 2010 | 66 | |
| AK011916.1 | - | 10 - 11 | 2048 | 66 | ||
| AK051027.1 | - | 10 - 11 | 2011 | 66 | ||
| BC053057.1 | - | 10 - 11 | 1987 | 5698028 | 66 | |
| BC086481.1 | - | 10 - 11 | 1994 | 6505546 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igdcc3
Genomic Map