Assay Details
| Gene Symbol: Dmrtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027732.2 | NP_082008.1 | 7 - 8 | 942 | 138 | |
| GenBank mRNA | AF542046.1 | - | 7 - 8 | 939 | 138 | |
| AK017020.1 | - | 7 - 8 | 940 | 138 | ||
| BC100434.1 | - | 7 - 8 | 942 | 30918218 | 138 | |
| BC138203.1 | - | 7 - 8 | 873 | 8861222 | 138 | |
| BC145988.1 | - | 7 - 8 | 873 | 40131291 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dmrtc2
Genomic Map