Assay Details
| Gene Symbol: Csnk1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173185.2 | NP_775277.2 | 4 - 5 | 646 | 132 | |
| GenBank mRNA | AK090262.1 | - | 4 - 5 | 647 | 132 | |
| AK149310.1 | - | 5 - 6 | 664 | 132 | ||
| BC064645.1 | - | 3 - 4 | 662 | 5363779 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Csnk1g1
Genomic Map