Assay Details
| Gene Symbol: Csnk1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173185.2 | NP_775277.2 | 9 - 10 | 1271 | 67 | |
| GenBank mRNA | AK090262.1 | - | 9 - 10 | 1272 | 67 | |
| BC064645.1 | - | 8 - 9 | 1287 | 5363779 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Csnk1g1
Genomic Map