Assay Details
Gene Symbol: 9230112J17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040464.1 | 2 - 3 | 224 | 66 | ||
GenBank mRNA | AK038653.1 | - | 2 - 3 | 227 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
thrombospondin, type I, domain containing 4 |
Thsd4 |
ADAMTSL-6, ADAMTSL6, AI585901, Adamtsl4, B230114P05Rik |
-4.2kb | Sense | Chr. 9 - 59966931 - 60522171 on NCBI Build GRCm38 |
Downstream | leucine rich repeat containing 49 | Lrrc49 |
AI430817, AW050057, D430025H09Rik, PGs4, p79 |
44.0kb | Sense | Chr. 9 - 60587229 - 60688134 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
