Assay Details
| Gene Symbol: Fgf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF020738.1 | - | 2 - 3 | 389 | 66 | |
| AK011712.2 | - | 2 - 3 | 488 | 66 | ||
| AK034335.1 | - | 2 - 3 | 347 | 66 | ||
| BB611389.1 | - | 3 - 4 | 354 | 66 | ||
| BC030485.1 | - | 3 - 4 | 311 | 5369273 | 66 | |
| CX239635.1 | - | 3 - 4 | 296 | 66 | ||
| U66201.1 | - | 2 - 3 | 310 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgf12
Genomic Map