Assay Details
| Gene Symbol: Fgf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF020738.1 | - | 3 - 4 | 497 | 114 | |
| AK011712.2 | - | 3 - 4 | 596 | 114 | ||
| AK034335.1 | - | 3 - 4 | 455 | 114 | ||
| BB611389.1 | - | 4 - 5 | 462 | 114 | ||
| BC030485.1 | - | 4 - 5 | 419 | 5369273 | 114 | |
| CX239635.1 | - | 4 - 5 | 404 | 114 | ||
| U66201.1 | - | 3 - 4 | 418 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgf12
Genomic Map