Assay Details
| Gene Symbol: Arhgap20os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033560.1 | 3 - 4 | 344 | 81 | ||
| GenBank mRNA | AK033819.1 | - | 3 - 4 | 263 | 81 | |
| AK135715.1 | - | 318 | 81 | |||
| AK165145.1 | - | 2 - 3 | 282 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK165145.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288541 | AK135715.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288541 | AK165145.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288541 | BG069615.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288541 | NR_033560.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288541 | AK033819.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288541 | AK135715.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288541 | AK165145.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288541 | BG069615.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288541 | NR_033560.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288541 | AK033819.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288541 | AK135715.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288541 | AK165145.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288541 | BG069615.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288541 | NR_033560.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288541 | AK033819.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288541 | AK135715.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288541 | AK165145.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288541 | BG069615.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288541 | NR_033560.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288541 | AK033819.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06788462 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06788452 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06788442 | - | - |
| Whole Mouse Genome G4122A | A_51_P288541 | AK135715.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288541 | AK165145.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288541 | BG069615.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288541 | NR_033560.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288541 | AK033819.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288541 | AK135715.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288541 | AK165145.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288541 | BG069615.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288541 | NR_033560.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288541 | AK033819.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133259 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06788452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07601531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26512004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08144777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07524367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07776483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26512003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08144778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07601530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08168857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07524350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07524354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26511925 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1456425_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10585286 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10593478 | - | - |
| Mouse Genome 430 2.0 Array | 1456425_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456425_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900019103.mm.1 | NR_033560.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019103.mm.1 | NR_033560.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019103.mm.1 | TC0900002290.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900019102.mm.1 | NR_033560.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019102.mm.1 | NR_033560.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019102.mm.1 | TC0900002290.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900019101.mm.1 | NR_033560.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019101.mm.1 | NR_033560.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019101.mm.1 | TC0900002290.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456425_at | - | - |
| Mouse Expression Set 430 | 1456425_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456425_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456425_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 400803 | - | - |

Arhgap20os
Genomic Map